missing translation for 'onlineSavingsMsg'
Learn More

Piceatannol, MedChemExpress

Product Code. 16431887
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16431887

Brand: Medchem Express HY13518

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Piceatannol is a well-known Syk inhibitor and reduces the expression of iNOS induced by TNF.

Piceatannol is an effective agent for research of acute lung injury (ALI). Piceatannol is a naturally occurring polyphenolic stilbene found in various fruits and vegetables and exhibits anticancer and anti-inflammatory properties. Piceatannol induces apoptosis in DLBCL cell lines. Piceatannol induces autophagy and apoptosis in MOLT-4 human leukemia cells.

Specifications

Chemical Name or Material Piceatannol
CAS 10083-24-6
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C14H12O4
Quantity 50 mg
Synonym Astringenin trans-Piceatannol
Solubility Information DMSO : ≥ 31 mg/mL (126.92 mM) ∣H2O : 1 mg/mL (4.09 mM; Need ultrasonic)
SMILES OC1=CC=C(/C=C/C2=CC(O)=CC(O)=C2)C=C1O
Molecular Weight (g/mol) 244.24
Formula Weight 244.24
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.