Learn More
Piceatannol, MedChemExpress
MedChemExpress Piceatannol is a well-known Syk inhibitor and reduces the expression of iNOS induced by TNF.
110.70 € - 376.65 €
Specifications
Chemical Name or Material | Piceatannol |
---|---|
CAS | 10083-24-6 |
Color | Earth Yellow |
Purity Grade Notes | Research |
Molecular Formula | C14H12O4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16443366
![]() |
Medchem Express
HY-13518 |
25 mg |
187.65 €
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16431887
![]() |
Medchem Express
HY-13518 |
50 mg |
264.60 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16416216
![]() |
Medchem Express
HY-13518 |
100 mg |
376.65 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16405728
![]() |
Medchem Express
HY-13518 |
10 mg |
110.70 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Piceatannol is an effective agent for research of acute lung injury (ALI). Piceatannol is a naturally occurring polyphenolic stilbene found in various fruits and vegetables and exhibits anticancer and anti-inflammatory properties. Piceatannol induces apoptosis in DLBCL cell lines. Piceatannol induces autophagy and apoptosis in MOLT-4 human leukemia cells.Specifications
Piceatannol | |
Earth Yellow | |
C14H12O4 | |
DMSO : ≥ 31 mg/mL (126.92 mM) ∣H2O : 1 mg/mL (4.09 mM; Need ultrasonic) | |
244.24 | |
98.09% | |
Solid |
10083-24-6 | |
Research | |
Astringenin trans-Piceatannol | |
OC1=CC=C(/C=C/C2=CC(O)=CC(O)=C2)C=C1O | |
244.24 | |
Research | |
Cancer-Kinase/protease |
Safety and Handling
H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.