missing translation for 'onlineSavingsMsg'
Learn More

Picosulfate Sodium, TRC

Product Code. 30137868
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137868 50 mg 50mg
30136513 10 mg 10mg
17924895 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137868 Supplier Toronto Research Chemicals Supplier No. TRCP43755050MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10040-45-6
Molecular Formula C18 H13 N O8 S2 . 2 Na
Molecular Weight (g/mol) 481.41
Synonym Sodium Picosulfate, Phenol, 4,4'-(2-pyridinylmethylene)bis-, 1,1'-bis(hydrogen sulfate), sodium salt (1:2), Phenol, 4,4'-(2-pyridinylmethylene)bis-, bis(hydrogen sulfate) (ester), disodium salt (9CI), Phenol, 4,4'-(2-pyridylmethylene)di-, bis(hydrogen sulfate) (ester) disodium salt (8CI), 4,4'-(2-Pyridylmethylene)diphenol bis(hydrogen sulfate) disodium salt, DA 1773, Disodium bis[4-[(hydroxysulfonyl)oxy]phenyl](2-pyridyl)methane, Evacuol, Guttalax, Guttalax-Fher, LA 391, Laxoberal, Laxoberon, Neopax, Pico-Salax, Picolax, Picosulfate sodium, Picosulfol, Rapilax
IUPAC Name disodium;[4-[pyridin-2-yl-(4-sulfonatooxyphenyl)methyl]phenyl] sulfate
SMILES [Na+].[Na+].[O-]S(=O)(=O)Oc1ccc(cc1)C(c2ccc(OS(=O)(=O)[O-])cc2)c3ccccn3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H15NO8S2.2Na/c20-28(21,22)26-15-8-4-13(5-9-15)18(17-3-1-2-12-19-17)14-6-10-16(11-7-14)27-29(23,24)25;;/h1-12,18H,(H,20,21,22)(H,23,24,25);;/q;2*+1/p-2
Formula Weight 480.9878
Chemical Name or Material Sodium Picosulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.