missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ 8-Well Polystyrene Strip Plates, Corner Notch

Product Code. 10309213 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 plates
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10309213 100 plates Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 10309213 Supplier Thermo Scientific™ Supplier No. 15031
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Pierce 8-Well Strip Plates are optically clear, polystyrene plates that have a standard 96-well microplate footprint but also allow individual 8-well strips (i.e., plate columns) to be removed from the rigid frame.

Additional Information and Online Ordering

The format provides convenience and quality-assurance when performing partial-plate assays because unused wells can be removed to prevent confusion and cross-contamination.

Highlights:

  • Optically clear polystyrene strip-well construction for compatibility with standard plate readers
  • High binding capacity surface improves the overall performance in immunoassay applications
  • Certified for performance, having consistently low coefficients of variation (CV) among wells
  • Eight-well strips fit securely in the plate frame, remaining in place even if turned upside down
  • Guides help to avoid misorientation of the strip when returned to the holder
  • Strips are gently removed from the frame with a simple twist or a strip-well ejector

One Strip Well Ejector per package.

 

 

 

TRUSTED_SUSTAINABILITY

Specifications

Binding Property Adsorption, Hydrophilic
Content And Storage Contents: 100 plates, 5 plates per sealed tray
Detection Method Colorimetric
Packaging 100 Plate
Product Line Pierce
Color Clear
Product Type Well Plate
Material Polystyrene
No. of Wells 96
Surface Treatment Polystyrene
Well Shape Flat Bottom (F-well)
Plate Blocking None
Quantity 100 plates
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.