missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ 9K MWCO Protein Concentrators

Product Code. 10003544 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 devices
25 devices
Unit Size:
Pack of 10
Pack of 25
This item is not returnable. View return policy

Product Code. 10003544

Brand: Thermo Scientific™ 87749

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Achieve high-performance concentration, diafiltration or buffer-exchange with these 9000-MW cut-off (MWCO) disposable centrifugal devices.

Thermo Scientific™ Pierce Protein Concentrators are disposable centrifugal devices for high-performance concentration, diafiltration or buffer-exchange of biological samples.

Pierce Protein Concentrators effectively combine simplicity, speed, capacity and recovery to yield enhanced concentration, purification and separation of proteins even with dilute protein samples. The high-performance regenerated cellulose membranes in these 9K molecular-weight cutoff (MWCO) units are carefully and accurately rated to provide more than 90% protein recovery for proteins larger than 9000 daltons. Pierce Concentrators are available in 7mL and 20mL sizes, which are compatible with swinging-bucket and fixed-angle rotors. These centrifugal concentrators provide efficient concentration of biological samples containing proteins (antigens, antibodies and enzymes), nucleic acids (DNA) and micro organisms (viral particles).

Highlights:

  • Rapid protein concentration – achieve greater than 100-fold protein concentration in less than 45 minutes

  • Excellent protein recovery – remove buffer and small molecules while retaining more than 90% of proteins larger than 9kDa

  • Convenient – two sizes designed to fit 15mL and 50mL conical tubes; ideal for simple concentration of 1 to 20mL of sample

  • Instrument-compatible – use standard centrifuges with swinging-bucket or fixed-angle rotors; collect and recover sample without invert spinning

  • Versatile – the easiest way to perform sample concentration, diafiltration or buffer-exchange of biological samples

Requires:

Centrifuge equipped with a rotor for 15mL or 50mL conical tubes (for 7mL or 20mL Concentrators, respectively)

Recommended for:

Protein concentration with tissue culture media, antiserum or monoclonal antibody preps; Concentration of protein peaks following gel-permeation chromatography; Removal of unincorporated protein label; Concentration and desalt/buffer-exchange after eluting protein from ion-exchange, hydrophobic interaction (HIC), metal chelate or affinity chromatography columns.

TRUSTED_SUSTAINABILITY

Specifications

Description Pierce Protein Concentrators, 9K MWCO, 20mL
Material (Membrane) Cellulose Membrane Bonded to Polypropylene Tube
MWCO 9 kDa
Quantity 10 devices
Sufficient For 10 samples, each 5 to 20mL
Volume (Metric) 20 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.