missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 96-Well Plates for Pierce™ BCA-RAC Assay

Product Code. 10166393 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 plates
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10166393 5 plates Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 10166393 Supplier Thermo Scientific™ Supplier No. 15045
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Reducing agent-compatible version of our popular BCA Reagent Kit to measure protein concentration in samples containing DTT or BME thiol-reductants.

This reducing agent-compatible (RAC) version of the Thermo Scientific™ Pierce BCA Protein Assay provides accurate determination of protein concentration in samples that contain dithiothreitol (DTT), 2-mercaptoethanol (BME), TCEP and other disulfide reducing agents.

The BCA Protein Assay Kit - Reducing Agent Compatible (BCA-RAC) provides all of the advantages of the original BCA Assay, plus compatibility with disulfide reducing agents at concentrations routinely used in protein sample buffers. This special adaptation of the popular Pierce BCA Protein Assay method enables accurate protein concentration measurement for samples containing DTT, 2-ME or TCEP. Available in tube and microplate formats, these reducing agent compatible (RAC) BCA Kits extend the already broad reagent compatibility of the BCA Protein Assay protocol to include nearly all types of components commonly present in protein research samples.

Highlights:

  • Compatible – assay samples that contain up to 5mM DTT, 35mM BME, or 10mM TCEP

  • BCA technology – only a slight modification of the standard BCA Protein Assay protocol (15-minute incubation with Compatibility Reagent); no precipitation steps required

  • Small samples – requires only 25μL (standard kit) or less than 10μL (microplate kit) of sample

  • Colorimetric – measure with a standard spectrophotometer or plate reader (562nm)

  • Excellent uniformity – exhibits less protein-to-protein variation than dye-binding methods

  • High linearity – linear working range for BSA equals 125 to 2000μg/mL

Includes:

BCA Reagents A and B, Compatibility Reagent, Reconstitution Buffer, and Albumin Standard (2mg/mL); Microplate kit also includes assay-compatible 96-well microplates

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Description Pierce 96-Well Microplates for BCA-RAC Assays
Detection Method Colorimetric
Packaging 5/Pk.
Product Line Pierce
Color Clear
Product Type Microplate
For Use With (Equipment) Spectrophotometer
Material Polystyrene
No. of Wells 96
Well Shape Round Bottom (U-well)
Quantity 5 plates
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.