missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Concentrated Buffer Stocks (10X and 20X)

Product Code. 11861465 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Concentration:
10X
20X
Chemical Name or Material:
Borate Buffer
DPBS
DPBS Tween 20 Buffer
PBS Tween 20 Buffer
Phosphate Buffered Saline (PBS)
TAE Buffer
TBE Buffer
TBS Buffer
TBS Tween 20 Buffer
Tris-Glycine Buffer
Tris-Glycine SDS Buffer
Unit Size:
1 L
500mL
Product Code. Quantity Concentration Chemical Name or Material unitSize
11861465 500 mL 20X TAE Buffer 500mL
11851465 500 mL 20X PBS Tween 20 Buffer 500mL
11881465 500 mL 20X TBS Tween 20 Buffer 500mL
10433985 500 mL 20X TBS Buffer 500mL
11831465 500 mL 20X Phosphate Buffered Saline (PBS) 500mL
10240444 1 L 10X Tris-Glycine SDS Buffer 1 L
10638294 500 mL 20X Borate Buffer 500mL
10405145 500 mL 20X DPBS 500mL
10493415 1 L 10X Tris-Glycine Buffer 1 L
11885115 1 L 10X TBE Buffer 1 L
15793722 500 mL 20X DPBS Tween 20 Buffer 500mL
11 options
This item is not returnable. View return policy

Product Code. 11861465

Brand: Thermo Scientific™ 28354

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Save time using Pierce Concentrated Buffer stock solutions (10X and 20X) with simple dilutions to expedite your experiment without sacrificing quality or consistency.

Our variety of Pierce Concentrated Buffer stock solutions are ready to use without having to weigh and dissolve dry ingredients or adjust the pH with concentrated acid or base. Simply dilute the liquid stock solution with pure water and proceed with your experiment.

Benefits of using Pierce Concentrated Buffer stock solutions include:
Convenient—no weighing, no pH adjustment, no need to stock individual components
Improved accuracy—ready-to-use formula, easily diluted without weighing chemical components
Consistent quality—our quality control ensures that every prep will yield the same, consistent buffer

20X PBS Tween-20 can be used to prepare PBS-Tween (PBS-T) wash buffers for ELISA, western, and other immunoassays, as well as blocking buffer for plate-based assays. When diluted to 1X with water, 20X PBS Tween-20 makes 10 mM sodium phosphate, 0.15 M NaCl, 0.05% Tween™ 20 buffer at pH 7.5.

20X TBS Tween-20 (Tris-buffered saline with Tween™ 20 detergent) can be used to prepare TBS-Tween (TBS-T) wash buffers or blocking buffer diluent for applications such as ELISA, western blotting, and other immunoassays. When diluted 20-fold in water, a TBS Tween-20 buffer containing 25 mM Tris, 0.15 M NaCl, 0.05% Tween-20 at pH 7.5 is ready for use.

20X Tris-Buffered Saline (TBS) is a stock solution for preparing Tris-NaCl buffer for use as a wash buffer and antibody diluent for ELISA, western blotting, and other immunoassays. 20X Tris Buffered Saline (TBS) makes 25 mM Tris, 0.15 M NaCl with pH 7.2 to 7.5 when diluted to 1X with water.

20X Phosphate-Buffered Saline (PBS) is ideal for preparing PBS buffers for use in crosslinking, biotinylation, and fluorescent labeling reactions requiring an amine-free buffer. When diluted 20-fold in water, the solution yields 10 mM sodium phosphate, 0.15 M NaCl at pH 7.5.

10X Tris-Glycine-SDS Buffer makes 0.025 M Tris, 0.192M glycine, 0.1% sodium dodecyl sulfate (SDS) at pH 8.5 when diluted to 1X with water.

20X Borate Buffer is ideal for preparing sodium borate buffer solutions for use in protein modification procedures requiring amine-free buffer at an alkaline pH. 20X Borate Buffer makes 50 mM borate at pH 8.8 when diluted to 1X with water.

20X Modified Dulbecco's PBS is used for preparing physiological Na- and K-phosphate buffered saline (D-PBS) used for wash buffers and antibody diluents in immunoassays like ELISA and western blot. 20X Modified Dulbecco's PBS makes 8 mM sodium phosphate, 2 mM potassium phosphate, 0.14 M NaCl, 10 mM KCl, pH 7.4 when diluted to 1X with water.

10X Tris-Glycine Buffer is a space-saving stock solution that is ideal for quickly preparing standard Tris-glycine (pH 8.5) transfer buffer used for western blotting using Tris-glycine gel electrophoresis. When diluted 10-fold in water or 20% methanol, the solution yields 0.025 M Tris, 0.192 M glycine at pH 8.5.

10X TBE Buffer stock solution is ideal for casting and preparing Tris-borate-EDTA (TBE) running buffer used for agarose and polyacrylamide gel electrophoresis of nucleic acids. When diluted to 1X, the TBE Buffer contains 0.089 M Tris, 0.089 M borate, 2 mM EDTA with pH 8.2 to 8.4.

20X Modified Dulbecco's PBS Tween-20 will make physiological sodium- and potassium-phosphate buffered saline (D-PBS) to be used as a wash buffer for ELISA, western, and other immunoassays or as diluent or blocking buffer for plate-based assays. 20X Modified Dulbecco's PBS Tween-20 (D-PBS-T) makes 8 mM sodium phosphate, 2 mM potassium phosphate, 0.14 M NaCl, 10 mM KCl, 0.05% Tween™ 20 at pH 7.4 when diluted to 1X with water.

20X TAE Buffer is a space-saving stock solution that is ideal for casting and preparing Tris-acetate EDTA (TAE) running buffer for agarose gel electrophoresis of nucleic acids. When diluted to 1X with water, a 0.04 M Tris, 0.04 M acetate, 1 mM EDTA buffer at pH 8.2 to 8.4 is ready for use.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material TAE Buffer
Concentration 20X
pH 8.3
Product Line Pierce
Quantity 500 mL
Formulation Acetic Acid
compliance-icons
Product Identifier
  • TAE Buffer
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : trometamol, acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.