missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Streptavidin Coated High Sensitivity Plates, Black, 8-Well Strip

Product Code. 11854514 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 plates
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 11854514

Brand: Thermo Scientific™ 15525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biotin-binding microplates that are coated to maximize capture efficiency of biotinylated probes and control background in assays for low levels of target analyte.

Thermo Scientific™ Pierce High Sensitivity Streptavidin Plates are biotin-binding microplates that are coated to maximize capture efficiency of biotinylated probes and control background in assays for low levels of target analyte.

Thermo Scientific Pierce Streptavidin Coated High Sensitivity Plates,Black, 8-Well Strip are biotin-binding microplates that are coated to maximize capture efficiency of biotinylated probes and control background in assays for low levels of target analyte.

Features of Streptavidin Coated High Sensitivity Plates, Black, 8-Well Strip

Sensitive—detect down to 5 ng/mL of biotinylated IgG
Specific—low background with high signal-to-noise ratios
Robust—broad dynamic range
Ready to use—plates are supplied pre-blocked to save time

Thermo Scientific Pierce Coated Plates are coated using proprietary surface-activation technology to achieve maximum efficient coating of active streptavidin biotin-binding protein. The high sensitivity plates enable detection of low concentrations (ng/mL) of biotinylated antibodies and other proteins.

Streptavidin is the most popular and widely available biotin-binding protein for research methods. Streptavidin has an isoelectric point of 5 to 6, resulting in low non-specific interactions.

Related Products
Pierce Streptavidin Coated High Sensitivity Plates, Clear, 8-Well Strip (Cat. No. 15520)

TRUSTED_SUSTAINABILITY

Specifications

Binding Property High Sensitivity, Affinity
Content And Storage Upon receipt store at 4°C. Product is shipped at ambient temperature.
Description Pierce Streptavidin Coated High Sensitivity Plates, Black, 96-Well Strip
Detection Method Fluorescence
Packaging 5 Plate
Product Line Pierce
Color Black
Product Type Microplate
Material Polystyrene
No. of Wells 96
Surface Treatment Streptavidin
Plate Blocking Blocker™ BSA Buffer
Quantity 5 plates
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.