missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Maleic Anhydride Activated Plates

Product Code. 10677764 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Clear
White
Format:
Standard
Strip Plate
Quantity:
5 Plates
5 x 5 Plates
Unit Size:
Pack of 25
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Format Quantity Unit Size
10677764 Clear Strip Plate 5 x 5 Plates Pack of 25
10503544 Clear Strip Plate 5 Plates Pack of 5
10115383 Clear Standard 5 Plates Pack of 5
10712794 Clear Standard 5 x 5 Plates Pack of 25
10379253 White Standard 5 Plates Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 10677764 Supplier Thermo Scientific™ Supplier No. 15102
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Thermo Scientific™ Pierce Maleic Anhydride Activated Plates allow attachment of amine-containing peptides and other molecules to microplate wells for use in binding assays.

Pierce Maleic Anhydride Activated Plates are useful for immobilizing peptides and other ligands that do not coat efficiently by passive adsorption. Reaction of the maleic anhydride groups with primary amines (–NH2) results in formation of amide bonds that are stable at neutral pH and above. Acidic conditions hydrolyze the bond; therefore, coating reactions are best performed at pH 8 to 9, and the reacted plates are best used at pH greater than 7 for ELISA and other methods.

These Thermo Scientific Pierce Maleic Anhydride Activated 96-Well Plates are clear, 96-well plates that allow attachment of amine-containing peptides and other molecules to microplate wells for use in binding assays.

Features of Maleic Anhydride Activated Plates:
• Spontaneously react with primary amines
• A direct, simple approach to covalently attach molecules to the well surface of an EIA plate
• Maleic anhydride maintains its integrity and availability for coupling for months under dry, room temperature conditions

TRUSTED_SUSTAINABILITY

Specifications

Binding Property Standard Capacity, Covalent
Format Strip Plate
Content And Storage Upon receipt store plates at room temperature. Product shipped at ambient temperature.
Detection Method Colorimetric
Product Line Pierce
Color Clear
Product Type Detection Plate, Immunoassay, ELISA
Material Polystyrene
No. of Wells 96
Surface Treatment Maleic Anhydride
No. per Pack 5 Plates
Plate Blocking None
Quantity 5 x 5 Plates
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.