missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Maleimide Activated Plates

Product Code. 10475015 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Black
Clear
White
Format:
Standard
Strip Plate
Detection Method:
Chemiluminescence
Colorimetric
Fluorescence
Unit Size:
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Format Detection Method unitSize
10475015 Black Standard Fluorescence Pack of 5
10328733 Clear Strip Plate Colorimetric Pack of 5
10348643 White Standard Chemiluminescence Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10475015 Supplier Thermo Scientific™ Supplier No. 15153

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Pierce Maleimide Activated Plates are ideal for binding sulfhydryl-containing molecules that are difficult to coat onto polystyrene plates, such as peptides that contain a terminal cysteine.

Pierce Maleimide Activated Plates provide covalent attachment (plate capture) of sulfhydryl-containing molecules, such as cysteine-terminated peptides. These coated plates are an especially useful tool for assessing specific anti-hapten antibody titers during antibody production. Maleimide Activated Plates are available in clear for colorimetric assays, white for chemiluminescent assays and black for fluorescent assays.

Highlights
• Spontaneous immobilization of cysteine-containin peptides and proteins with free sulfhydryls
• Pre-blocked to reduce nonspecific binding

TRUSTED_SUSTAINABILITY

Specifications

Binding Property Standard Capacity, Covalent
Format Standard
Content And Storage Note: Plates are supplied pre-blocked with bovine serum albumin.

Upon receipt store plates at 4°C protected from moisture. Once opened, place unused plates in a resealable bag with desiccant and store at 4°C. Plates are shipped at ambient temperature.
Detection Method Fluorescence
Product Line Pierce
Color Black
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 100 μL
No. of Wells 96
Surface Treatment Maleimide
No. per Pack 5 Plates
Plate Blocking Blocker™ BSA Buffer
Quantity 5 plates
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.