Learn More
PIEZO1 Antibody (CL9714) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88938JF525
Product Code. 30292966
  • 737.00 € / 0.10mL
Description
PIEZO1 Monoclonal antibody specifically detects PIEZO1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PIEZO1
Monoclonal
Janelia Fluor 525
FAM38A, member A, membrane protein induced by beta-amyloid treatment, Mib, piezo-type mechanosensitive ion channel component 1
This antibody was developed using a synthetic peptide derived from Q92508, with the exact immunogen sequence remaining proprietary.
0.1 mL
Alzheimers Research, Neuroscience
9780
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL9714
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified