missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIEZO2 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78538H
This item is not returnable.
View return policy
Description
PIEZO2 Polyclonal specifically detects PIEZO2 in Human, Mouse, Porcine samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PIEZO2 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Q9H5I5 | |
| PIEZO2 | |
| A synthetic peptide made to an internal portion of the human PIEZO2 protein (between residues 1450-1500). [UniProt Q9H5I5] | |
| 0.1 mL | |
| Alzheimers Research, Neuroscience | |
| 63895 | |
| Human, Mouse, Pig | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| PBS with No Preservative | |
| C18orf30, C18orf58, FAM38B, FAM38B2, family with sequence similarity 38, member B, family with sequence similarity 38, member B2, FLJ23144, FLJ23403, FLJ25916, FLJ34907, FLJ37734, FLJ45725, HsT748, HsT771, piezo-type mechanosensitive ion channel component 2, protein PIEZO2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and mouse. Porcine reactivity reported in scientific literature (PMID: 25385580). | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction