missing translation for 'onlineSavingsMsg'
Learn More

PIEZO2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

379.00 € - 529.00 €

Specifications

Antigen PIEZO2
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18425322
View Documents
Novus Biologicals
NBP1-89893-25ul
25 μL
379.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18421802
View Documents
Novus Biologicals
NBP1-89893
0.1 mL
529.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PIEZO2 Polyclonal antibody specifically detects PIEZO2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

PIEZO2
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Alzheimers Research, Neuroscience
PBS (pH 7.2) and 40% Glycerol
63895
IgG
Immunogen affinity purified
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Purified
RUO
Human
C18orf30, C18orf58, FAM38B, FAM38B2, family with sequence similarity 38, member B, family with sequence similarity 38, member B2, FLJ23144, FLJ23403, FLJ25916, FLJ34907, FLJ37734, FLJ45725, HsT748, HsT771, piezo-type mechanosensitive ion channel component 2, protein PIEZO2
This antibody was developed against Recombinant Protein corresponding to amino acids: ESEKPIFTMSAQQSQLKVMDQQSFNKFIQAFSRDTGAMQFLENYEKEDITVAELEGNSNSLWTISPPSKQKMIHELLDPNSSFSVVFSWSIQRNLSLGAKSEIATDKLSFPLKNITRKNIAKM
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.