missing translation for 'onlineSavingsMsg'
Learn More

Pig anti-Human IgA/IgG/IgM (H+L) Secondary Antibody, R&D Systems™

Product Code. 18168024 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18168024 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18168024 Supplier Novus Biologicals Supplier No. NBP269258
  • ONLINE EXCLUSIVE PRICE
    472.67 €
    418.95 € / 10mg
    Save 53.72 €
    11% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Pig Polyclonal Antibody

IgA/IgG/IgM (H+L) Polyclonal specifically detects IgA/IgG/IgM (H+L) in Human samples. It is validated for Immunoelectrophoresis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgA/IgG/IgM (H+L)
Applications Immunoelectrophoresis
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Radioimmunodiffusion, Immunoelectrophoresis
Formulation Lyophilized from PBS (pH 7.2) with No Preservative
Host Species Pig
Immunogen Highly purified IgG and pools of homogenous IgA and IgM isolated from human serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Ion exchange chromatography
Quantity 10 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity The reactivity of the antiserum is directed to the major isotypes of the human immunoglobulin system (classes and both light chain types) including antibodies to common determinants, to class and to the surface determinants of the common Fab portion, as tested by immunoelectrophoresis and double radial immunodiffusion against pooled serum and purified immunoglobulins. In immunoelectrophoresis and double radial immunodiffusion using various antiserum concentrations against normal human plasma and serum, the characteristic IgG, IgA and IgM precipitin lines are obtained.
Reconstitution Reconstitute by by adding 1 ml sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Lyophilized
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.