missing translation for 'onlineSavingsMsg'
Learn More

Pig anti-Human IgG (H+L) Secondary Antibody, Biotin, R&D Systems™

Product Code. 18118593 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18118593 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18118593 Supplier Novus Biologicals Supplier No. NBP269398
  • ONLINE EXCLUSIVE PRICE
    487.00 €
    425.25 € / 1mL
    Save 61.75 €
    12% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Pig Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Human samples. It is validated for Dot Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Dot Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
Formulation Lyophilized from PBS (pH 7.2) with No Preservative
Host Species Pig
Immunogen Purified normal IgG isolated from pooled human serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Ion exchange chromatography
Quantity 1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity The reactivity of the antiserum is directed to the Fc and Fab subunits of the IgG molecule. It includes a certain degree of reactivity with other immunoglobulins via the common Fab portion. It does not react with any non-Ig protein in human serum, as tested by immunoelectrophoresis and double radial immunodiffusion.
Reconstitution Reconstitute by by adding 1 ml sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Lyophilized
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.