missing translation for 'onlineSavingsMsg'
Learn More

Pig anti-Human Kappa Light Chain Secondary Antibody, FITC, R&D Systems™

Product Code. 18176652 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18176652 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18176652 Supplier Novus Biologicals Supplier No. NBP269239
  • ONLINE EXCLUSIVE PRICE
    553.50 €
    511.35 € / 1mL
    Save 42.15 €
    7% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Pig Polyclonal Antibody

Kappa Light Chain Polyclonal specifically detects Kappa Light Chain in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kappa Light Chain
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration LYOPH
Conjugate FITC
Dilution ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
Formulation Lyophilized from PBS (pH 7.2) with No Preservative
Gene Alias HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809
Gene Symbols IGKC
Host Species Pig
Immunogen A pool of purified Bence Jones kappa proteins isolated from human urine. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Ion exchange chromatography
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3514
Test Specificity The reactivity of the antiserum is directed to the surface and hidden determinants of Ig kappa light chain. In immunoelectrophoresis this antiserum is reacting with polyclonal and monoclonal immunoglobulins of the kappa type, Bence Jones proteins as well as free light chains of the kappa type. This antiserum does not react with any other protein of human serum or plasma.
Reconstitution Reconstitute by by adding 1 ml sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Lyophilized
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.