missing translation for 'onlineSavingsMsg'
Learn More

Pig anti-Rabbit IgG (H+L) Secondary Antibody, R&D Systems™

Product Code. 18631799 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18631799 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18631799 Supplier Novus Biologicals Supplier No. NBP269422
  • ONLINE EXCLUSIVE PRICE
    352.98 €
    306.60 € / 1mL
    Save 46.38 €
    13% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Pig Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for Immunoelectrophoresis
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Immunoelectrophoresis
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Formulation Whole antiserum
Host Species Pig
Immunogen Purified normal IgG isolated from pooled rabbit serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Whole Antiserum
Quantity 1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Reconstitution Reconstitute by adding 1 mL sterile distilled water. Working dilutions are prepared by adding sterile phosphate buffered saline (PBS, pH 7.2).
Target Species Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Lyophilized
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.