missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIGU Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 624.00 €
Specifications
| Antigen | PIGU |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18491762
|
Novus Biologicals
NBP2-30386-25ul |
25ul |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18109114
|
Novus Biologicals
NBP2-30386 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PIGU Polyclonal specifically detects PIGU in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PIGU | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| bA346K17.2, CDC91 (cell division cycle 91, S. cerevisiae, homolog)-like 1, CDC91L1CDC91 cell division cycle 91-like 1 (S. cerevisiae), cell division cycle 91-like 1 protein, Cell division cycle protein 91-like 1, GAB1, GPI transamidase component PIG-U, GPI transamidase subunit, MGC40420, phosphatidylinositol glycan anchor biosynthesis class U protein, phosphatidylinositol glycan anchor biosynthesis, class U, Protein CDC91-like 1 | |
| PIGU | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q9H490 | |
| 128869 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KLLLELDQYAPDVAELIRTPMEMRYIPLKVALFYLLNPYTILSCVAKSTCAINN | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title