Learn More
PIK3CA Antibody (CL8345) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-05502MFV450
Product Code. 30286694
  • 759.00 € / 0.10mL
Description
PIK3CA Monoclonal antibody specifically detects PIK3CA in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PIK3CA
Monoclonal
mFluor Violet 450 SE
EC 2.7.1, EC 2.7.1.153, MGC142161, MGC142163, p110-alpha, Phosphatidylinositol 3-Kinase catalytic subunit alpha, phosphatidylinositol 3-kinase, catalytic, 110-KD, alpha, phosphatidylinositol 3-kinase, catalytic, alpha polypeptide, Phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha isoform, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit, alpha isoform, phosphoinositide-3-kinase, catalytic, alpha polypeptide, PI3K, PI3K-alpha, PI3-kinase p110 subunit alpha, PI3-kinase subunit alpha, PtdIns-3-kinase p110, PtdIns-3-kinase subunit alpha, PtdIns-3-kinase subunit p110-alpha
This antibody was developed using a synthetic peptide derived from P42336, with the exact immunogen sequence remaining proprietary.
0.1 mL
Breast Cancer, Cancer, Diabetes Research, mTOR Pathway, Oncogenes
5290
Store at 4⪚C in the dark.
IgG1
Western Blot
CL8345
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified