missing translation for 'onlineSavingsMsg'
Learn More

Pilaralisib analogue, MedChemExpress

Product Code. 16454398
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16454398 10 mg 10mg
16450717 50 mg 50mg
16415296 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16454398 Supplier Medchem Express Supplier No. HY11105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pilaralisib analogue (XL147 analogue) is a representative and selective PI3Kα inhibitor extracted from patent WO2012006552A1, Compound 147 in Table 1.

Pilaralisib analogue (XL147 analogue) is a representative and selective PI3Kα inhibitor extracted from patent WO2012006552A1, Compound 147 in Table 1.

Specifications

Chemical Name or Material Pilaralisib analogue
CAS 956958-53-5
Color Brown
Purity Grade Notes Research
Molecular Formula C21H16N6O2S2
Quantity 10 mg
Synonym XL147 analogue
Solubility Information DMSO : ≥ 50 mg/mL (111.48 mM)
SMILES O=S(C1=CC=C(C)C=C1)(NC2=NC3=CC=CC=C3N=C2NC4=CC5=NSN=C5C=C4)=O
Molecular Weight (g/mol) 448.52
Formula Weight 448.52
Percent Purity 99.67%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.