missing translation for 'onlineSavingsMsg'
Learn More

Pimecrolimus, MedChemExpress

Product Code. 16441977
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16441977 50 mg 50mg
16415828 10 mg 10mg
16426296 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16441977 Supplier Medchem Express Supplier No. HY13723
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Pimecrolimus is an immunophilin ligand, which binds specifically to the cytosolic receptor, immunophilin macrophilin-12.

Pimecrolimus is an immunophilin ligand, which binds specifically to the cytosolic receptor, immunophilin macrophilin-12.

Specifications

Chemical Name or Material Pimecrolimus
Physical Form Solid
CAS 137071-32-0
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C43H68ClNO11
For Use With (Application) COVID-19-immunoregulation
Synonym SDZ-ASM 981
Solubility Information DMSO : ≥ 32 mg/mL (39.48 mM)
SMILES O=C([C@@](CCCC1)([H])N1C(C([C@@]2(O)[C@H](C)C[C@H](OC)[C@@](O2)([H])[C@@H](OC)C[C@@H](C)C/C(C)=C/[C@H]3CC)=O)=O)O[C@H](/C(C)=C/[C@H]4C[C@@H](OC)[C@@H](Cl)CC4)[C@H](C)[C@@H](O)CC3=O
Molecular Weight (g/mol) 810.45
Formula Weight 810.45
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.