missing translation for 'onlineSavingsMsg'
Learn More

Pinaverium bromide, MedChemExpress

Product Code. 16480807
Encompass_Preferred
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 16480807

Brand: Medchem Express HY111613

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pinaverium bromide is an L-type calcium channel blocker with selectivity for the gastrointestinal tract, effectively relieves pain, diarrhea and intestinal discomfort, provides good therapeutic efficacies without significant adverse effects on Irritable bowel syndrome (IBS) patients.

Pinaverium bromide is an L-type calcium channel blocker with selectivity for the gastrointestinal tract, effectively relieves pain, diarrhea and intestinal discomfort, provides good therapeutic efficacies without significant adverse effects on Irritable bowel syndrome (IBS) patients.

Specifications

Chemical Name or Material Pinaverium bromide
CAS 53251-94-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H41Br2NO4
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (84.54 mM; Need ultrasonic)
SMILES CC1(C)C2CCC(CCOCC[N+]3(CC4=CC(OC)=C(OC)C=C4Br)CCOCC3)C1C2.[Br-]
Molecular Weight (g/mol) 591.42
Formula Weight 591.42
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.