missing translation for 'onlineSavingsMsg'
Learn More

Pioglitazone, MedChemExpress

Product Code. 16405938
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16405938 10 mg 10mg
16402087 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16405938 Supplier Medchem Express Supplier No. HY13956

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pioglitazone (U 72107) is an orally active and selective PPARγ (peroxisome proliferator-activated receptor) agonist with high affinity binding to the PPARγ ligand-binding domain with EC50 of 0.93 and 0.99 μM for human and mouse PPARγ, respectively. Pioglitazone can be used in diabetes research.

Pioglitazone (U 72107) is an orally active and selective PPARγ (peroxisome proliferator-activated receptor) agonist with high affinity binding to the PPARγ ligand-binding domain with EC50 of 0.93 and 0.99 μM for human and mouse PPARγ, respectively. Pioglitazone can be used in diabetes research.

Specifications

Chemical Name or Material Pioglitazone
CAS 111025-46-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H20N2O3S
Quantity 10 mg
Synonym U 72107
Solubility Information DMSO : 40 mg/mL (112.22 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(N1)SC(CC2=CC=C(OCCC3=NC=C(CC)C=C3)C=C2)C1=O
Molecular Weight (g/mol) 356.44
Formula Weight 356.44
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.