missing translation for 'onlineSavingsMsg'
Learn More

Pipemidic acid, MedChemExpress

Product Code. 16438686
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16438686

Brand: Medchem Express HYB1210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pipemidic acid, a derivative of Piromidic acid, is an antibacterial agent. Pipemidic acid is active against gram-negative bacteria including Pseudomonas aeruginosa as well as some gram-positive bacteria. Pipemidic acid can be used for the research of intestinal, urinary, and biliary tract infections.

Pipemidic acid, a derivative of Piromidic acid, is an antibacterial agent. Pipemidic acid is active against gram-negative bacteria including Pseudomonas aeruginosa as well as some gram-positive bacteria. Pipemidic acid can be used for the research of intestinal, urinary, and biliary tract infections.

Specifications

Chemical Name or Material Pipemidic acid
Physical Form Solid
CAS 51940-44-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C14H17N5O3
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 5 mg/mL (16.48 mM; Need ultrasonic)
SMILES O=C(C1=CN(CC)C2=NC(N3CCNCC3)=NC=C2C1=O)O
Molecular Weight (g/mol) 303.32
Formula Weight 303.32
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.