missing translation for 'onlineSavingsMsg'
Learn More

Piperacetazine, MedChemExpress

Product Code. 16468678
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468678 10 mg 10mg
16461098 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16468678 Supplier Medchem Express Supplier No. HYB1152
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Piperacetazine is an antipsychotic prodrug, used forschizophrenia.

Piperacetazine is an antipsychotic prodrug, used forschizophrenia.

Specifications

Chemical Name or Material Piperacetazine
CAS 3819-00-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H30N2O2S
Quantity 10 mg
Solubility Information Ethanol : 50 mg/mL (121.78 mM; Need ultrasonic) ∣DMSO : ≥ 6 mg/mL (14.61 mM)
SMILES CC(C(C=C1N2CCCN3CCC(CCO)CC3)=CC=C1SC4=C2C=CC=C4)=O
Molecular Weight (g/mol) 410.57
Formula Weight 410.57
Percent Purity 99.03%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.