missing translation for 'onlineSavingsMsg'
Learn More

Piperidino(3-piperidinyl)methanone hydrochloride, 90%, Thermo Scientific™

Product Code. 10350703
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10350703 250 mg Amber Glass Bottle 250mg
10366352 1 g Amber Glass Bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10350703

Brand: Thermo Scientific Maybridge MO01165CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 40576-21-4
Molecular Formula C11H20N2O
Molecular Weight (g/mol) 196.294
MDL Number MFCD01580193
InChI Key OSZRYTYCKGZYLB-UHFFFAOYSA-N
Synonym 1-piperidin-3-ylcarbonyl piperidine, piperidin-1-yl piperidin-3-yl methanone, piperidino 3-piperidyl ketone, 1,3'-carbonyldi piperidine, ketone, piperidino 3-piperidyl, 3-piperidinocarbonyl piperidine, 1-piperidine-3-carbonyl piperidine, piperidine, 1-3-piperidyl carbonyl, 3-piperidine-1-carbonyl piperidine, piperidin-3-yl-piperidin-1-yl-methanone
PubChem CID 218422
IUPAC Name piperidin-1-yl(piperidin-3-yl)methanone
SMILES C1CCN(CC1)C(=O)C2CCCNC2

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 232.75
Percent Purity 90%
Chemical Name or Material Piperidino(3-piperidinyl)methanone hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.