missing translation for 'onlineSavingsMsg'
Learn More

Piperidolate, MedChemExpress

Product Code. 16491446
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491446 200 mg 200mg
16468536 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491446 Supplier Medchem Express Supplier No. HYB0962A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Piperidolate is an antimuscarinic, inhibits intestinal cramp induced by acetylcholine (rats and dogs).

Piperidolate is an antimuscarinic, inhibits intestinal cramp induced by acetylcholine (rats and dogs).

Specifications

Chemical Name or Material Piperidolate
Physical Form Viscous Liquid
CAS 82-98-4
Color Colorless
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C21H25NO2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 150 mg/mL (463.78 mM)
SMILES O=C(OC1CN(CC)CCC1)C(C2=CC=CC=C2)C3=CC=CC=C3
Molecular Weight (g/mol) 323.43
Formula Weight 323.43
Percent Purity 99.34%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.