missing translation for 'onlineSavingsMsg'
Learn More

Piperine, MedChemExpress

Product Code. 16451318
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
Unit Size:
1g
200mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451318 5 g 5g
16492499 1 g 1g
16411496 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16451318 Supplier Medchem Express Supplier No. HYN0144
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Piperine, a natural alkaloid isolated from Piper nigrum L, inhibits P-glycoprotein and CYP3A4 activities with an IC50 value of 61.94±0.054 μg/mL in HeLa cell.

Piperine, a natural alkaloid isolated from Piper nigrum L, inhibits P-glycoprotein and CYP3A4 activities with an IC50 value of 61.94±0.054 μg/mL in HeLa cell.

Specifications

Chemical Name or Material Piperine
CAS 94-62-2
Color White
Purity Grade Notes Research
Molecular Formula C17H19NO3
Quantity 5 g
Synonym Bioperine 1-Piperoylpiperidine
Solubility Information DMSO : 50 mg/mL (175.23 mM; Need ultrasonic)
SMILES O=C(N1CCCCC1)/C=C/C=C/C2=CC=C(OCO3)C3=C2
Molecular Weight (g/mol) 285.34
Formula Weight 285.34
Percent Purity 98.88%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.