missing translation for 'onlineSavingsMsg'
Learn More

Piperine, MedChemExpress

Product Code. p-200096014
Encompass_Preferred
Click to view available options
Quantity:
200 mg
1 g
5 g
Unit Size:
1g
200mg
5g
This item is not returnable. View return policy

Product Code. 16492499

Brand: Medchem Express HYN0144

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Piperine, a natural alkaloid isolated from Piper nigrum L, inhibits P-glycoprotein and CYP3A4 activities with an IC50 value of 61.94±0.054 μg/mL in HeLa cell.

Piperine, a natural alkaloid isolated from Piper nigrum L, inhibits P-glycoprotein and CYP3A4 activities with an IC50 value of 61.94±0.054 μg/mL in HeLa cell.

Specifications

Chemical Name or Material Piperine
CAS 94-62-2
Color White
Purity Grade Notes Research
Molecular Formula C17H19NO3
Quantity 1 g
Synonym Bioperine 1-Piperoylpiperidine
Solubility Information DMSO : 50 mg/mL (175.23 mM; Need ultrasonic)
SMILES O=C(N1CCCCC1)/C=C/C=C/C2=CC=C(OCO3)C3=C2
Molecular Weight (g/mol) 285.34
Formula Weight 285.34
Percent Purity 98.88%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.