missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Piperonyl isobutyrate, 99%

Product Code. 11419237
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419237 25 g 25g
11429237 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419237 Supplier Alfa Aesar™ Supplier No. B21321.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5461-08-5
MDL Number MFCD00005826

Specifications

Density 1.154
Boiling Point 91°C to 92°C (0.005 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C12H9ClN2O4S
Refractive Index 1.512
Quantity 25 g
Synonym n-2-chlorophenyl-3-nitrobenzenesulfonamide, 2'-chloro-3-nitrobenzenesulfonanilide, cambridge id 5138232, 2-chlorophenyl 3-nitrophenyl sulfonyl amine, n-2-chlorophenyl-3-nitrobenzene-1-sulfonamide, n-2-chlorophenyl-3-nitrobenzenesulfonamide #
InChI Key LQIWGYRYMZKQQL-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)NS(=O)(=O)C2=CC=CC(=C2)[N+](=O)[O-])Cl
IUPAC Name N-(2-chlorophenyl)-3-nitrobenzenesulfonamide
Molecular Weight (g/mol) 312.72
PubChem CID 232034
Formula Weight 222.24
Percent Purity 99%
Chemical Name or Material Piperonyl isobutyrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.