missing translation for 'onlineSavingsMsg'
Learn More

Pirarubicin, MedChemExpress

Product Code. 16451977
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451977 50 mg 50mg
16436296 100 mg 100mg
16435828 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16451977 Supplier Medchem Express Supplier No. HY13725
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pirarubicin is an anthracycline antibiotics, acts as a topoisomerase II inhibitor, and is a widely used for treatment of various cancers, in particular, solid tumors.

Pirarubicin is an anthracycline antibiotics, acts as a topoisomerase II inhibitor, and is a widely used for treatment of various cancers, in particular, solid tumors.

Specifications

Chemical Name or Material Pirarubicin
Physical Form Solid
CAS 72496-41-4
Color Red
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C32H37NO12
For Use With (Application) Cancer-programmed cell death
Synonym THP
Solubility Information DMSO : 10 mg/mL (15.93 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C1C2=C(C=CC=C2OC)C(C3=C(O)C4=C([C@@H](O[C@@]5([H])C[C@H](N)[C@H](O[C@@]6([H])OCCCC6)[C@H](C)O5)C[C@@](C(CO)=O)(O)C4)C(O)=C31)=O
Molecular Weight (g/mol) 627.64
Formula Weight 627.64
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.