missing translation for 'onlineSavingsMsg'
Learn More

Pirfenidone, MedChemExpress

Product Code. 16442196
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16442196 500 mg 500mg
16432259 1 g 1g
16418416 100 mg 100mg
16451038 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16442196 Supplier Medchem Express Supplier No. HYB0673
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Pirfenidone (AMR69) is an antifibrotic agent that attenuates CCL2 and CCL12 production in fibrocyte cells. Pirfenidone has growth-inhibitory effect and reduces TGF-β2 protein levels in human glioma cell lines. Pirfenidone also has anti-inflammatory activities.

Pirfenidone (AMR69) is an antifibrotic agent that attenuates CCL2 and CCL12 production in fibrocyte cells. Pirfenidone has growth-inhibitory effect and reduces TGF-β2 protein levels in human glioma cell lines. Pirfenidone also has anti-inflammatory activities.

Specifications

Chemical Name or Material Pirfenidone
Physical Form Powder
CAS 53179-13-8
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C12H11NO
For Use With (Application) COVID-19-immunoregulation
Synonym AMR69
Solubility Information DMSO : ≥ 100 mg/mL (539.90 mM) ∣H2O : 16.67 mg/mL (90.00 mM; Need ultrasonic)
SMILES O=C1C=CC(C)=CN1C2=CC=CC=C2
Molecular Weight (g/mol) 185.22
Formula Weight 185.22
Percent Purity 99.15%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.