missing translation for 'onlineSavingsMsg'
Learn More

Pirimiphos-methyl, 90%, TRC

Product Code. 30133547
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133547 10 g 10g
18040050 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133547 Supplier Toronto Research Chemicals Supplier No. TRCP50878010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29232-93-7
Molecular Formula C11 H20 N3 O3 P S
Molecular Weight (g/mol) 305.33
Synonym 4-Pyrimidinol, 2-(diethylamino)-6-methyl-, O-ester with O,O-dimethyl phosphorothioate (8CI), 2-Diethylamino-6-methylpyrimidin-4-yl dimethyl phosphorothionate, 2-Diethylamino-6-methylpyrimidine-4-yl dimethyl phosphorothionate, Actellic, Actellic 25EC, Actellic 50, Actellic 50E, Actellic 50EC, Actellic Polvo, Actellic Powder, Blex, Dominator, ENT 27699Gc, Methyl pyrimiphos, Methylpirimiphos, Methylpyrimifos, O-[2-(Diethylamino)-6-methyl-4-pyrimidinyl] O,O-dimethyl phosphorothioate, Orafon, PP 511, Pirimiphos M, Pirimiphos Me, Pirimiphos methyl, Piritione, Plant Protection PP 511, Pyrimidine phosphate, Pyrimiphos methyl, Rotator, Silosan, Tomahawk, VUCHT 388
IUPAC Name 4-dimethoxyphosphinothioyloxy-N,N-diethyl-6-methylpyrimidin-2-amine
SMILES CCN(CC)c1nc(C)cc(OP(=S)(OC)OC)n1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H20N3O3PS/c1-6-14(7-2)11-12-9(3)8-10(13-11)17-18(19,15-4)16-5/h8H,6-7H2,1-5H3
Formula Weight 305.0963
Chemical Name or Material Pirimiphos-methyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.