missing translation for 'onlineSavingsMsg'
Learn More

Piroxicam, MedChemExpress

Product Code. 16472109
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472109 1 g 1g
16468438 10 g 10g
16450848 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16472109 Supplier Medchem Express Supplier No. HYB0253
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Piroxicam (CP-16171) is a non-steroidal anti-inflammatory drugs, acts as a COX inhibitor, with IC50s of 47, 25 μM for human monocyte COX-1 and COX-2, respectively.

Piroxicam (CP-16171) is a non-steroidal anti-inflammatory drugs, acts as a COX inhibitor, with IC50s of 47, 25 μM for human monocyte COX-1 and COX-2, respectively.

Specifications

Chemical Name or Material Piroxicam
CAS 36322-90-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C15H13N3O4S
Quantity 1 g
Synonym CP-16171
Solubility Information DMSO : 50 mg/mL (150.90 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=C(O)C2=CC=CC=C2S(N1C)(=O)=O)NC3=NC=CC=C3
Molecular Weight (g/mol) 331.35
Formula Weight 331.35
Percent Purity 99.61%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.