missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PITX3 Antibody (SR1536), Novus Biologicals™
Rabbit Monoclonal Antibody
349.00 € - 449.00 €
Specifications
| Antigen | PITX3 |
|---|---|
| Clone | SR1536 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunoprecipitation 1:50 |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18683954
|
Novus Biologicals
NBP3-22365-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18612285
|
Novus Biologicals
NBP3-22365-50ul |
50 μg |
349.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PITX3 Monoclonal antibody specifically detects PITX3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunoprecipitationSpecifications
| PITX3 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CTPP4, Homeobox protein PITX3, MGC12766, paired-like homeodomain 3, Paired-like homeodomain transcription factor 3, pituitary homeobox 3, PTX3 | |
| A synthesized peptide derived from human PITX3 (Uniprot #: O75364) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1536 | |
| Western Blot, Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Neurodegeneration, Neuroscience | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 5309 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title