Learn More
Invitrogen™ PIWIL1 Recombinant Rabbit Monoclonal Antibody (24GB1545)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557607
 View more versions of this product
Product Code. 30275506
  • 198.00 € / 20µL
Description
PIWIL1 Recombinant Monoclonal Antibody for Western Blot, Flow
Hiwi is a member of the Piwi /Argonaute protein family. Hiwi is specifically expressed in germline cells such as spermatocytes and round spermatids during spermatogenesis. Hiwi is also expressed in CD34+ hematopoietic progenitors, but not in CD34- hematopoietic cells, suggesting a role for this gene in the maintenance of hematopoietic stem cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PIWIL1
Recombinant Monoclonal
Unconjugated
PIWIL1
CT80; CT80.1; HILI; HIWI; mili; Miwi; Miwi like; Miwi likw; PIWI; piwi homolog; piwi like homolog 1; piwi like RNA-mediated gene silencing 1; PIWIL1; PIWIL1L; piwil2-like protein; piwi-like 1; piwi-like homolog 1; piwi-like protein 1; piwi-like protein 2; piwi-like RNA-mediated gene silencing 1
Rabbit
Affinity chromatography
RUO
363912, 57749, 9271
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1545
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q96J94, Q9JMB7
PIWIL1
A synthesized peptide derived from human PIWIL1 (aa 754-800).
20 μL
Primary
Human, Mouse, Rat