missing translation for 'onlineSavingsMsg'
Learn More

Pizotifen, MedChemExpress

Product Code. 16461376
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461376 200 mg 200mg
16458106 100 mg 100mg
16412026 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16461376 Supplier Medchem Express Supplier No. HYB0115
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pizotifen (Pizotyline) is a potent 5-HT2 receptor antagonist, with a high affinity for 5-HT1C binding site.

Pizotifen (Pizotyline) is a potent 5-HT2 receptor antagonist, with a high affinity for 5-HT1C binding site.

Specifications

Chemical Name or Material Pizotifen
Physical Form Solid
CAS 15574-96-6
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C19H21NS
For Use With (Application) Neuroscience-Neuromodulation
Synonym Pizotyline BC-105
Solubility Information DMSO : 20 mg/mL (67.70 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CN1CC/C(CC1)=C2C3=CC=CC=C3CCC4=C\2C=CS4
Molecular Weight (g/mol) 295.44
Formula Weight 295.44
Percent Purity 99.73%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.