Learn More
Invitrogen™ PKC alpha/beta/gamma Recombinant Rabbit Monoclonal Antibody (24GB885)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557174
 View more versions of this product
Product Code. 30275425
  • 146.00 € / 20µL
Description
PKC alpha/beta/gamma Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Protein Kinase c alpha (PKC alpha) is an 77 kDa member of the conventional group (cPKCs: sensitive to calcium, diacylglycerol, phosphatidylserine and phorbol esters) of the PKC family of serine/ threonine kinases that are involved in a wide range of physiological processes including mitogenesis, cell survival and transcriptional regulation. PKC alpha is an ubiquitously expressed PKC isozyme that has been implicated in the regulation of a broad range of cellular functions including proliferation, differentiation, development, migration, cell cell adhesion, cell extracellular matrix adhesion, and solute transport. The activation loop threonine (threonine 497 in PKC alpha) of conventional PKCs is phosphorylated by phosphoinositide dependent kinase 1 (PDK1). This phosphorylation is necessary for the autophosphorylation of threonine 638 in the carboxy terminus of PKC alpha, a step that is critical for regulating the rate of PKC alpha dephosphorylation and inactivation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PKC alpha/beta/gamma
Recombinant Monoclonal
2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P05129, P05132, P05771, P17252, P27791, P63318, P63319, P68403, P68404
PRKACA, PRKCB, PRKCG
A synthesized peptide derived from human PKC (aa 300-500).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB885
Unconjugated
PRKACA
A130082F03Rik; cAMP-dependent protein kinase catalytic subunit alpha; cAMP-dependent protein kinase catalytic subunit alpha, isoform 1; cAMP-dependent protein kinase catalytic subunit C alpha; Cs-PKA; PKA C-alpha; Pkaca; PKC; PKCA1; Pkcb; PKC-B; PKC-beta; PKCC; PKCD; PKCG; PKCgamma; PKC-gamma; PKCI; PPNAD4; PRKACA; Prkc; Prkcb; Prkcb1; Prkcb2; Prkcc; Prkcg; protein kinase A; protein kinase A alpha; protein kinase A catalytic subunit; protein kinase C beta; protein kinase C beta I; protein kinase C beta II; protein kinase C beta type; protein kinase C beta-II; protein kinase C gamma; protein kinase C gamma type; protein kinase C type I (gamma type); protein kinase C, beta; protein kinase C, beta 1; protein kinase C, beta 1 polypeptide; protein kinase C, gamma; protein kinase cAMP-activated catalytic subunit alpha; protein kinase, cAMP dependent, catalytic, alpha; protein kinase, cAMP-dependent, alpha catalytic subunit; protein kinase, cAMP-dependent, catalytic, alpha; RATPKCI; SCA14; sperm cAMP-dependent protein kinase catalytic subunit
Rabbit
Affinity chromatography
RUO
18747, 18751, 18752, 24681, 25023, 25636, 5566, 5579, 5582
-20°C