missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC delta, p Ser299 Antibody (S06-4A5), Novus Biologicals™
Rabbit Monoclonal Antibody
421.00 € - 645.00 €
Specifications
| Antigen | PKC delta |
|---|---|
| Clone | S06-4A5 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18638963
|
Novus Biologicals
NBP3-19827-100ul |
100 μg |
645.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18675073
|
Novus Biologicals
NBP3-19827-50ul |
50 μg |
421.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PKC delta Monoclonal antibody specifically detects PKC delta in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| PKC delta | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific, Protein Kinase, Signal Transduction | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 5580 | |
| IgG | |
| Affinity purified |
| S06-4A5 | |
| Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 2.7.11, EC 2.7.11.13, MAY1, MGC49908, nPKC-delta, PKCD, PKCy, protein kinase C delta type, protein kinase C delta VIII, protein kinase C, delta | |
| A synthetic phosphopeptide corresponding to residues surrounding Ser299 of human PKC delta (Uniprot # Q05655) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title