missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PKC delta Recombinant Rabbit Monoclonal Antibody (24GB4485)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557646
This item is not returnable.
View return policy
Description
PKC delta Recombinant Monoclonal Antibody for Western Blot, Flow
The PKC family of serine/threonine kinases, including PRKCD (PKC delta), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II, and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. Protein kinase C (PKC) can be activated by calcium and the second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play distinct roles in cells. The protein encoded by this gene is one of the PKC family members. Studies both in human and mice demonstrate that this kinase is involved in B cell signaling and in the regulation of growth, apoptosis, and differentiation of a variety of cell types. Alternatively spliced transcript variants encoding the same protein have been observed.
Specifications
| PKC delta | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PRKCD | |
| 1-May; AI385711; ALPS3; CVID9; D14Ertd420e; kinase nPKC-delta; KPCD; MAY1; nPKC-delta; PKC[d]; Pkcd; PKCdelta; Prkcd; protein kinase C delta; protein kinase C delta type; Protein kinase C delta type catalytic subunit; Protein kinase C delta type regulatory subunit; protein kinase C delta variant IX; protein kinase C delta VIII; protein kinase C, delta; protein kinase C, delta IV; protein kinase C, delta V; protein kinase C[d]; SDK1; Sphingosine-dependent protein kinase-1; Tyrosine-protein kinase PRKCD | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 170538, 18753, 5580 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 24GB4485 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P09215, P28867, Q05655 | |
| PRKCD | |
| Recombinant protein of Mouse PKC delta (aa 540-674). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction