missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PKC delta/theta Recombinant Rabbit Monoclonal Antibody (23GB4585), Invitrogen™
Description
PRKCD (Protein Kinase C Delta) is a Protein Coding gene. Diseases associated with PRKCD includeAutoimmune Lymphoproliferative Syndrome, Type IiiandSystemic Lupus Erythematosus 16. Among its related pathways areRole of Calcineurin dependent NFAT signaling in lymphocytesandWnt Signaling Pathway and Pluripotency. Gene Ontology (GO) annotations related to this gene includetransferase activity, transferring phosphorus containing groupsandprotein tyrosine kinase activity. An important paralog of this gene isPRKCQ (PKC theta).
Specifications
Specifications
| Antigen | PKC delta/theta |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 23GB4585 |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | PRKCQ |
| Gene Accession No. | P09215, P28867, Q02111, Q04759, Q05655, Q9WTQ0 |
| Gene Alias | A130035A12Rik; AI385711; ALPS3; AW494342; CVID9; D14Ertd420e; may1; npkc-delta; nPKC-theta; PKC theta; PKC[d]; PKC-0; Pkcd; PKCd1; PKCd2; PKCdelta; PKC-delta; PKCdelta1; pkc-delta1; PKCdelta2; Pkcq; PKCtheta; PKC-theta; Prkcd; prkcd.L; prkcd.S; prkcd-a; prkcd-b; PRKCQ; PRKCT; protein kinase C delta; protein kinase C delta S homeolog; protein kinase C delta type; Protein kinase C delta type catalytic subunit; Protein kinase C delta type regulatory subunit; protein kinase C delta variant IX; protein kinase C delta VIII; protein kinase C theta; protein kinase C theta type; protein kinase C, delta; protein kinase C, delta IV; protein kinase C, delta L homeolog; protein kinase C, delta V; protein kinase C, theta; protein kinase C[d]; protein kinase delta1; SDK1; SDK-1; Sphingosine-dependent protein kinase-1; Tyrosine-protein kinase PRKCD; XELAEV_18025733mg |
| Gene Symbols | PRKCD, PRKCQ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?