missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKR Antibody (6H3A10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
196.00 € - 593.00 €
Specifications
| Antigen | PKR |
|---|---|
| Clone | 6H3A10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18037383
|
Novus Biologicals
NBP2-37242 |
0.1 mL |
593.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18693226
|
Novus Biologicals
NBP2-37242-0.025ml |
0.025 mL |
196.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PKR Monoclonal specifically detects PKR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| PKR | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| double stranded RNA activated protein kinase, EC 2.7.11.1, eIF-2A protein kinase 2, EIF2AK1, eukaryotic translation initiation factor 2-alpha kinase 2P1/eIF-2A protein kinase, interferon-induced, double-stranded RNA-activated protein kinase, interferon-inducible double stranded RNA dependent, interferon-inducible elF2alpha kinase, Interferon-inducible RNA-dependent protein kinase, PKRp68 kinase, PRKRMGC126524, Protein kinase RNA-activated | |
| EIF2AK2 | |
| IgG1 | |
| Affinity Purified |
| 6H3A10 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Apoptosis, Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| 5610 | |
| Purified recombinant fragment of human EIF2AK2 (AA: 329-551) expressed in E. Coli. | |
| Primary | |
| 62 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title