missing translation for 'onlineSavingsMsg'
Learn More

PKR Antibody (S04-3F2), Novus Biologicals™

Rabbit Monoclonal Antibody

421.00 € - 645.00 €

Specifications

Antigen PKR
Clone S04-3F2
Concentration 0.3 mg/mL
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18657824
View Documents
Novus Biologicals
NBP3-19538-100ul
100 μg
645.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18695014
View Documents
Novus Biologicals
NBP3-19538-50ul
50 μg
421.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PKR Monoclonal antibody specifically detects PKR in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications

Specifications

PKR
0.3 mg/mL
Monoclonal
Purified
RUO
Human, Mouse, Rat
double stranded RNA activated protein kinase, EC 2.7.11.1, eIF-2A protein kinase 2, EIF2AK1, eukaryotic translation initiation factor 2-alpha kinase 2P1/eIF-2A protein kinase, interferon-induced, double-stranded RNA-activated protein kinase, interferon-inducible double stranded RNA dependent, interferon-inducible elF2alpha kinase, Interferon-inducible RNA-dependent protein kinase, PKRp68 kinase, PRKRMGC126524, Protein kinase RNA-activated
Recombinant protein of mouse PKR (Uniprot # Q03963)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
S04-3F2
Western Blot, ELISA
Unconjugated
Rabbit
Apoptosis, Phospho Specific, Signal Transduction
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
5610
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.