missing translation for 'onlineSavingsMsg'
Learn More

PLA2G16 Monoclonal Antibody (OTI1B6), TrueMAB™, OriGene

Product Code. 17546562
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17546562 30 μL 30µL
17526562 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17546562 Supplier Origene Technologies Supplier No. TA506911S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PLA2G16 Monoclonal antibody specifically detects PLA2G16 in Human samples. It is validated for Western Blot

Specifications

Antigen PLA2G16
Applications Western Blot
Classification Monoclonal
Clone OTI1B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PLAAT3
Gene Accession No. P53816
Gene Alias Adipose-specific phospholipase A2; adipose-specific PLA2; AdPLA; C78643; Ca-independent phospholipase A1/2; group XVI phospholipase A1/A2; Group XVI phospholipase A2; HRAS like suppressor 3; HRAS-like suppressor 1; HRAS-like suppressor 3; HRASLS3; H-rev 107 protein; H-rev 107 protein homolog; Hrev107; H-REV107; H-REV107-1; HREV107-1; HREV107-3; HRSL3; LRAT-like protein 3; MLP-3; Phospholipase A and acyltransferase 3; phospholipase A2 group XVI; phospholipase A2, group XVI; Pla2g16; PLAAT3; Renal carcinoma antigen NY-REN-65
Host Species Mouse
Immunogen Full length human recombinant protein of human PLA2G16 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11145
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.