missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2R1 Antibody (PLA2R1/4898), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44600-100ug
This item is not returnable.
View return policy
Description
PLA2R1 Monoclonal antibody specifically detects PLA2R1 in Human samples. It is validated for Immunohistochemistry
Specifications
| PLA2R1 | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 22925 | |
| Store at 4⪚C. | |
| IgG1 κ |
| Immunohistochemistry | |
| PLA2R1/4898 | |
| Unconjugated | |
| 180 kDa secretory phospholipase A2 receptor, CLEC13CC-type lectin domain family 13 member C, phospholipase A2 receptor 1, 180kDa, PLA2G1R, PLA2IR, PLA2-RM-type receptor, PLA2Rphospholipase A2 receptor 1, 180kD, secretory phospholipase A2 receptor | |
| Recombinant fragment (around aa300-600) of human PLA2R1 protein (Uniprot: Q13018). Exact sequence is proprietary. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction