missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLAC4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
168.00 € - 507.00 €
Specifications
| Antigen | PLAC4 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663156
|
Novus Biologicals
NBP2-41125-0.025mg |
0.025 mg |
168.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18120318
|
Novus Biologicals
NBP2-41125 |
0.1 mg |
507.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLAC4 Polyclonal specifically detects PLAC4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| PLAC4 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 191585 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the center of human PLAC4. The immunogen is located within amino acids 20 - 70 of PLAC4. Amino Acid Squence: DTLHPRHRHSSPWTPH | |
| Primary |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| C21orf115, Chromosome 21 Open Reading Frame 115, D21S418E, Placenta-Specific 4, Placenta-Specific Protein 4, PRED78 | |
| PLAC4 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title