missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLAGL1 Antibody (8D8C5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
196.00 € - 537.00 €
Specifications
| Antigen | PLAGL1 |
|---|---|
| Clone | 8D8C5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18798863
|
Novus Biologicals
NBP2-37343 |
0.1 mL |
537.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18693746
|
Novus Biologicals
NBP2-37343-0.025ml |
0.025 mL |
196.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLAGL1 Monoclonal specifically detects PLAGL1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PLAGL1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| DKFZp781P1017, LOT-1, LOT1ZACLost on transformation 1, MGC126275, MGC126276, PLAG-like 1, pleiomorphic adenoma gene-like 1, pleiomorphic adenoma gene-like protein 1, Pleiomorphic adenoma-like protein 1, Tumor supressor ZAC, ZAC tumor supressor, ZAC1, zinc finger protein PLAGL1 | |
| PLAGL1 | |
| IgG1 | |
| Affinity Purified |
| 8D8C5 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Mouse | |
| Apoptosis, Neuronal Stem Cell Markers, Stem Cell Markers | |
| PBS with 0.05% Sodium Azide | |
| 5325 | |
| Purified recombinant fragment of human PLAGL1 (AA: 118-222) expressed in E. Coli. | |
| Primary | |
| 50.8 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title