missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plakophilin 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 572.00 €
Specifications
| Antigen | Plakophilin 1 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18468271
|
Novus Biologicals
NBP1-90041-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18230866
|
Novus Biologicals
NBP1-90041 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Plakophilin 1 Polyclonal specifically detects Plakophilin 1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Plakophilin 1 | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| B6PBand 6 protein, MGC138829, plakophilin 1 (ectodermal dysplasia/skin fragility syndrome), plakophilin-1 | |
| PKP1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Cellular Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 5317 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RFSSYSQMENWSRHYPRGSCNTTGAGSDICFMQKIKASRSEPDLYCDPRGTLRKGTLGSKGQKTTQNRYSFYSTCSGQKAIKKCPVRPPSCASKQDPV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title