missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plakophilin 2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94257-0.02ml
This item is not returnable.
View return policy
Description
Plakophilin 2 Polyclonal antibody specifically detects Plakophilin 2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Plakophilin 2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ARVD9, MGC177501, plakophilin 2, plakophilin-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 578-837 of human PKP2 (NP_001005242.2). LPEKYSQNIYIQNRNIQTDNNKSIGCFGSRSRKVKEQYQDVPMPEEKSNPKGVEWLWHSIVIRMYLSLIAKSVRNYTQEASLGALQNLTAGSGPMPTSVAQTVVQKESGLQHTRKMLHVGDPSVKKTAISLLRNLSRNLSLQNEIAKETLPDLVSIIPDTVPSTDLLIETTASACYTLNNIIQNSYQNARDLLNTGGIQKIMAISAGDAYASNKASKAASVLLYSLWAHTELHHAYKKAQFKKTDFVNSRTAKAYHSLKD | |
| 0.02 mL | |
| Angiogenesis, Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
| 5318 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction