missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Plasma Kallikrein Monoclonal Antibody (736811)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561161
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Plasma prekallikrein is a glycoprotein that participates in the surface-dependent activation of blood coagulation, fibrinolysis, kinin generation and inflammation. It is synthesized in the liver and secreted into the blood as a single polypeptide chain. Plasma prekallikrein is converted to plasma kallikrein by factor XIIa by the cleavage of an internal Arg-Ile bond. Plasma kallikrein therefore is composed of a heavy chain and a light chain held together by a disulphide bond. The heavy chain originates from the amino-terminal end of the zymogen and contains 4 tandem repeats of 90 or 91 amino acids. Each repeat harbors a novel structure called the apple domain. The heavy chain is required for the surface-dependent pro-coagulant activity of plasma kallikrein. The light chain contains the active site or catalytic domain of the enzyme and is homologous to the trypsin family of serine proteases. Plasma prekallikrein deficiency causes a prolonged activated partial thromboplastin time in patients.
Specifications
| Plasma Kallikrein | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P03952 | |
| Klkb1 | |
| NS0 mouse myeloma cell line transfected with human KLK-B1, Gly20-Ala638. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin) | |
| 736811 | |
| Unconjugated | |
| Klkb1 | |
| antigen, prostate specific; APS; EC 3.4.21.34; Fletcher factor; Kal3; Kal-3; kallikrein; kallikrein 3, plasma; kallikrein B plasma; kallikrein B, plasma (Fletcher factor) 1; kallikrein B, plasma 1; kallikrein B1; Kalp1; KALP15; kininogenin; Klk3; KLKB1; Pk; PKK; PKKD; plasma kallikrein; Plasma kallikrein heavy chain; Plasma kallikrein light chain; Plasma prekallikrein; PPK; prekallikrein; PSA | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3818 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction